Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myo1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161817.2 | NP_001155289.1 | 5 | 629 | ||
NM_001290982.1 | NP_001277911.1 | 5 | 629 | |||
NM_010863.4 | NP_034993.2 | 5 | 629 | |||
GenBank mRNA | AK138265.1 | - | 5 | 564 | ||
AK141207.1 | - | 5 | 604 | |||
AK149409.1 | - | 5 | 576 | |||
AK161062.1 | - | 5 | 593 | |||
AK189624.1 | - | 1 | 16 | |||
BC026826.1 | - | 5 | 577 | |||
BC046300.1 | - | 5 | 575 | 6487332 | ||
BC054786.1 | - | 5 | 579 | 6821232 | ||
L00923.1 | - | 5 | 598 | |||
X69987.1 | - | 5 | 549 |