Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010877.4 | NP_035007.1 | 1, 2 | 324 | ||
GenBank mRNA | AB002664.1 | - | 1, 2 | 278 | ||
AK036379.1 | - | 1, 2 | 325 | |||
AK137152.1 | - | 1, 2 | 379 | |||
AK153447.1 | - | 1, 2 | 286 | |||
AK155324.1 | - | 1, 2 | 379 | |||
AK170839.1 | - | 1, 2 | 349 | |||
AK170998.1 | - | 1, 2 | 352 | |||
AK171059.1 | - | 1, 2 | 350 | |||
AK171265.1 | - | 1, 2 | 352 | |||
BC003730.1 | - | 1, 2 | 272 | 3592460 | ||
BC076609.1 | - | 2, 3 | 1278 | |||
BC094056.1 | - | 1, 2 | 370 | |||
DQ449939.1 | - | 1, 2 | 279 |