Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nkx3-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010921.3 | NP_035051.1 | 2 | 446 | ||
GenBank mRNA | AK137281.1 | - | 2 | 457 | ||
BC050109.1 | - | 2 | 443 | |||
BC132401.1 | - | 2 | 500 | 40130678 | ||
BC132427.1 | - | 2 | 500 | 40130704 | ||
U73460.1 | - | 2 | 433 | |||
U88542.1 | - | 2 | 446 |