Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005417.4 | NP_005408.1 | 14 | 3888 | ||
NM_198291.2 | NP_938033.1 | 14 | 3813 | |||
XM_011529013.2 | XP_011527315.1 | 15 | 4157 | |||
XM_017028024.1 | XP_016883513.1 | 16 | 4181 | |||
XM_017028025.1 | XP_016883514.1 | 15 | 4062 | |||
XM_017028026.1 | XP_016883515.1 | 15 | 3901 | |||
XM_017028027.1 | XP_016883516.1 | 13 | 3478 | |||
GenBank mRNA | AK024281.1 | - | 7 | 3085 | ||
AK091756.1 | - | 3 | 2660 | |||
BC011566.1 | - | 12 | 3478 | 4548399 | ||
BC051270.1 | - | 12 | 3541 | 4871614 |