Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plod1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF046782.1 | - | 1, 2 | 110 | ||
BY312508.1 | - | 1, 2 | 182 |
Gene Symbol: Myo5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC057910.1 | - | 109 |