Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Numb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136075.2 | NP_001129547.1 | 3 | 229 | ||
NM_010949.2 | NP_035079.1 | 3 | 229 | |||
GenBank mRNA | AF169192.1 | - | 3 | 214 | ||
AF170709.1 | - | 3 | 214 | |||
AK164313.1 | - | 1 | 2383 | |||
BC050108.1 | - | 6 | 486 |