Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pax9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011041.2 | NP_035171.1 | 4 | 1443 | ||
GenBank mRNA | AK034985.1 | - | 4 | 1440 | ||
AK132404.1 | - | 4 | 1444 | |||
BC005794.1 | - | 4 | 1385 | 3707718 | ||
BC006930.1 | - | 3 | 2068 | |||
X84000.1 | - | 4 | 1417 |