Gene Symbol: Plaur | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011113.3 | NP_035243.1 | 4, 5 | 488 | ||
GenBank mRNA | AK002580.1 | - | 4, 5 | 559 | ||
AK131951.1 | - | 4, 5 | 504 | |||
AK144796.1 | - | 4, 5 | 505 | |||
AK146827.1 | - | 4, 5 | 503 | |||
AK154141.1 | - | 4, 5 | 505 | |||
AK155351.1 | - | 4, 5 | 492 | |||
X62700.1 | - | 4, 5 | 489 |