Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Numb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136075.2 | NP_001129547.1 | 6 | 600 | ||
NM_001272055.1 | NP_001258984.1 | 5 | 567 | |||
NM_001272056.1 | NP_001258985.1 | 7 | 797 | |||
NM_010949.2 | NP_035079.1 | 6 | 600 | |||
GenBank mRNA | AF169191.1 | - | 5 | 552 | ||
AF169192.1 | - | 6 | 585 | |||
AF170709.1 | - | 6 | 585 | |||
AK004553.1 | - | 8 | 822 | |||
BC033459.1 | - | 8 | 838 | 3991630 | ||
BC050108.1 | - | 9 | 857 | |||
U70674.1 | - | 7 | 718 |