Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ogg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010957.4 | NP_035087.3 | 7 | 1316 | ||
GenBank mRNA | AK146870.1 | - | 7 | 1316 | ||
AK165875.1 | - | 7 | 1269 | |||
AK218768.1 | - | 2 | 157 | |||
BC138733.1 | - | 7 | 1272 | 8861755 | ||
BC138736.1 | - | 7 | 1272 | 8861758 | ||
U88621.1 | - | 7 | 1284 | |||
U96711.1 | - | 7 | 1281 | |||
Y11247.1 | - | 7 | 1247 |