Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkcg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291434.1 | NP_001278363.1 | 13 | 1587 | ||
NM_011102.4 | NP_035232.1 | 14 | 1740 | |||
GenBank mRNA | AK144419.1 | - | 14 | 1742 | ||
BC111807.1 | - | 13 | 1474 | 40039740 | ||
L28035.1 | - | 14 | 1818 | |||
X67129.1 | - | 14 | 1549 |