Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prrx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025570.1 | NP_001020741.1 | 1 | 1110 | ||
NM_011127.2 | NP_035257.1 | 1 | 1110 | |||
NM_175686.3 | NP_783617.2 | 1 | 1110 | |||
GenBank mRNA | AB221686.1 | - | 1 | 95 | ||
AK029347.1 | - | 1 | 1131 | |||
AK031075.1 | - | 1 | 76 | |||
AK032773.1 | - | 1 | 677 | |||
AK084387.1 | - | 1 | 1128 | |||
AK089006.1 | - | 1 | 942 | |||
BC092372.1 | - | 1 | 459 | 30533016 | ||
L06502.1 | - | 1 | 275 | |||
U03873.1 | - | 1 | 1116 | |||
X59725.1 | - | 1 | 236 |