Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ppib | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011149.2 | NP_035279.2 | 1 | 219 | ||
GenBank mRNA | AK002357.1 | - | 1 | 166 | ||
AK128977.1 | - | 1 | 169 | |||
AK154073.1 | - | 1 | 158 | |||
AL363449.1 | - | 1 | 260 | |||
BC013061.1 | - | 1 | 156 | 3483267 | ||
M60456.1 | - | 1 | 138 | |||
X58990.1 | - | 1 | 264 |