Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lgals3bp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011150.2 | NP_035280.1 | 4 | 425 | ||
GenBank mRNA | AK079915.1 | - | 4 | 454 | ||
AK137836.1 | - | 4 | 402 | |||
AK150021.1 | - | 4 | 454 | |||
AK152182.1 | - | 4 | 367 | |||
AK153313.1 | - | 4 | 441 | |||
AK153396.1 | - | 4 | 443 | |||
AK155031.1 | - | 4 | 367 | |||
BC090658.1 | - | 4 | 421 | 30666981 | ||
L16894.1 | - | 4 | 432 | |||
X67809.1 | - | 4 | 425 |