Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmb7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011187.1 | NP_035317.1 | 2, 3 | 168 | ||
GenBank mRNA | AK002823.1 | - | 2, 3 | 183 | ||
AK012613.1 | - | 2, 3 | 169 | |||
AK013961.1 | - | 2, 3 | 179 | |||
AK031913.1 | - | 2, 3 | 184 | |||
AK075759.1 | - | 2, 3 | 184 | |||
AK088276.1 | - | 2, 3 | 179 | |||
AK088765.1 | - | 2, 3 | 182 | |||
AK162002.1 | - | 2, 3 | 187 | |||
AK168823.1 | - | 2, 3 | 171 | |||
BC033424.1 | - | 2, 3 | 165 | |||
BC049230.1 | - | 2, 3 | 161 | |||
BC057662.1 | - | 2, 3 | 169 | 2865594 | ||
D83585.1 | - | 2, 3 | 168 | |||
Y10874.1 | - | 2, 3 | 182 |