Gene Symbol: TAF15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003487.3 | NP_003478.1 | 15, 16 | 1849 | ||
NM_139215.2 | NP_631961.1 | 15, 16 | 1858 | |||
XM_011525315.1 | XP_011523617.1 | 15, 16 | 1828 | |||
XM_017025181.1 | XP_016880670.1 | 13, 14 | 1701 | |||
GenBank mRNA | AK304520.1 | - | 16, 17 | 1750 | ||
AK313223.1 | - | 15, 16 | 1815 | |||
AK314194.1 | - | 15, 16 | 1829 | |||
BC039129.1 | - | 1, 2 | 399 | |||
BC046099.1 | - | 15, 16 | 1790 | 6067044 | ||
BG403710.1 | - | 2, 3 | 610 | |||
U51334.1 | - | 15, 16 | 1829 | |||
X98893.1 | - | 15, 16 | 1780 |