Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptpre | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011212.3 | NP_035342.3 | 6, 7 | 739 | ||
GenBank mRNA | AK004602.1 | - | 6, 7 | 739 | ||
AK132455.1 | - | 5, 6 | 702 | |||
AK160947.1 | - | 3, 4 | 288 | |||
BC024631.1 | - | 6, 7 | 733 | |||
BC053408.1 | - | 6, 7 | 515 | |||
BC098473.1 | - | 6, 7 | 503 | |||
D83484.1 | - | 6, 7 | 1076 | |||
U35368.1 | - | 6, 7 | 739 | |||
U36758.1 | - | 3, 4 | 288 | |||
U40280.1 | - | 5, 6 | 479 | |||
U62387.1 | - | 6, 7 | 641 |