Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rasgrp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011246.2 | NP_035376.1 | 2 | 257 | ||
GenBank mRNA | AF106070.1 | - | 2 | 101 | ||
AK013548.1 | - | 2 | 303 | |||
AK141524.1 | - | 2 | 382 | |||
BC057120.1 | - | 2 | 257 | 6831137 | ||
BC057341.1 | - | 2 | 257 | 6831137 |