Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lgmn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011175.2 | NP_035305.1 | 4 | 399 | ||
GenBank mRNA | AF044266.1 | - | 4 | 398 | ||
AJ000990.1 | - | 4 | 399 | |||
AK148104.1 | - | 3 | 347 | |||
AK149666.1 | - | 4 | 454 | |||
AK180466.1 | - | 128 | ||||
BC132515.1 | - | 3 | 346 | 40130792 | ||
BC132519.1 | - | 3 | 346 | 40130796 |