Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptp4a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011200.2 | NP_035330.1 | 5 | 1076 | ||
GenBank mRNA | AK081491.1 | - | 4 | 542 | ||
AK148288.1 | - | 5 | 1072 | |||
AK150506.1 | - | 4 | 706 | |||
AK150556.1 | - | 3 | 1659 | |||
AK150762.1 | - | 3 | 1660 | |||
AK151003.1 | - | 3 | 1660 | |||
AK151533.1 | - | 4 | 706 | |||
BC003761.1 | - | 5 | 953 | |||
BC031734.1 | - | 5 | 998 | |||
BC055039.1 | - | 5 | 1032 | 6396041 | ||
BC086787.1 | - | 4 | 382 | 30538771 | ||
BC094447.1 | - | 5 | 943 | 3990529 | ||
BC150972.1 | - | 4 | 748 | 9087885 | ||
BC150981.1 | - | 4 | 751 | 9087894 | ||
U84411.1 | - | 5 | 1085 |