Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PSAP SIRNA Assays ›
Gene Symbol:
Psap
Gene Name:
prosaposin
Gene Aliases:
AI037048, SGP-1
Chromosome Location:
Chr.10: 60277628 - 60302600 on Build GRCm38
UniGene ID:
Mm.277498
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
68967
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Psap
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001146120.1
NP_001139592.1
9
1044
NM_001146121.1
NP_001139593.1
10
1050
NM_001146122.1
NP_001139594.1
9
1041
NM_001146123.1
NP_001139595.1
9
1017
NM_001146124.1
NP_001139596.1
9
960
NM_011179.3
NP_035309.3
10
1053
GenBank mRNA
AK088369.1
-
9
958
AK089998.1
-
9
962
AK093881.1
-
9
958
AK135637.1
-
9
958
AK145607.1
-
9
965
AK145802.1
-
9
959
AK148152.1
-
9
985
AK148545.1
-
10
969
AK149785.1
-
9
961
AK150331.1
-
9
960
AK150645.1
-
9
958
AK151034.1
-
9
958
AK151173.1
-
9
959
AK151230.1
-
9
958
AK151234.1
-
9
933
AK151251.1
-
9
931
AK151525.1
-
9
960
AK151529.1
-
9
933
AK151686.1
-
9
994
AK151852.1
-
9
960
AK151875.1
-
9
957
AK151891.1
-
9
958
AK151907.1
-
9
960
AK151943.1
-
9
960
AK152117.1
-
9
960
AK152126.1
-
9
958
AK152134.1
-
9
960
AK152203.1
-
9
992
AK152280.1
-
9
958
AK152319.1
-
9
962
AK152355.1
-
9
960
AK152408.1
-
9
963
AK152505.1
-
9
960
AK152557.1
-
9
960
AK152605.1
-
9
960
AK152819.1
-
9
972
AK153404.1
-
9
958
AK159108.1
-
9
988
AK159164.1
-
9
958
AK159167.1
-
9
959
AK159198.1
-
9
958
AK159221.1
-
9
959
AK159244.1
-
9
955
AK159266.1
-
9
958
AK159333.1
-
9
958
AK159341.1
-
9
958
AK159384.1
-
9
958
AK159414.1
-
9
961
AK159427.1
-
9
960
AK159451.1
-
9
958
AK159457.1
-
9
972
AK159463.1
-
9
960
AK159487.1
-
9
958
AK159492.1
-
9
955
AK159510.1
-
9
958
AK159528.1
-
9
958
AK159530.1
-
9
958
AK159589.1
-
9
960
AK159604.1
-
9
958
AK159622.1
-
8
850
AK159634.1
-
9
958
AK159638.1
-
9
959
AK159657.1
-
9
958
AK159664.1
-
9
958
AK159685.1
-
9
958
AK159692.1
-
9
958
AK159709.1
-
9
958
AK159720.1
-
9
958
AK159722.1
-
9
958
AK159728.1
-
9
960
AK159840.1
-
9
958
AK159861.1
-
9
958
AK159862.1
-
9
960
AK159877.1
-
9
958
AK159910.1
-
9
958
AK159915.1
-
9
958
AK159936.1
-
9
960
AK159982.1
-
9
958
AK159994.1
-
9
958
AK160015.1
-
9
958
AK160019.1
-
9
960
AK160021.1
-
9
958
AK160024.1
-
9
958
AK160046.1
-
9
958
AK160049.1
-
9
960
AK160073.1
-
9
958
AK160108.1
-
9
958
AK160877.1
-
9
961
AK161313.1
-
9
975
AK166700.1
-
9
958
AK166707.1
-
9
957
AK166744.1
-
9
956
AK166811.1
-
9
958
AK166849.1
-
9
958
AK166879.1
-
9
960
AK166909.1
-
9
958
AK166920.1
-
9
958
AK166946.1
-
9
963
AK166959.1
-
9
958
AK166984.1
-
9
958
AK167014.1
-
9
958
AK167025.1
-
9
958
AK167073.1
-
9
958
AK167099.1
-
9
961
AK167108.1
-
9
958
AK167111.1
-
9
958
AK167147.1
-
9
958
AK167198.1
-
9
965
AK167213.1
-
9
958
AK167237.1
-
9
958
AK167243.1
-
9
959
AK167252.1
-
9
960
AK167418.1
-
9
960
AK167719.1
-
9
960
AK167902.1
-
9
1028
AK170416.1
-
9
957
AK182197.1
-
3
195
AK191456.1
-
3
201
AK195069.1
-
3
201
AK196371.1
-
3
201
BC036266.1
-
8
824
BC141089.1
-
10
963
9055701
BC145359.1
-
9
957
9053917
S36200.1
-
10
943
S71616.1
-
10
983
U27340.1
-
9
958
Back To Top
More Information
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Gene Ontology Categories:
Function(s)
Process(es)
lipid metabolic process
sphingolipid metabolic process
adenylate cyclase-inhibiting G-protein coupled receptor signaling pathway
regulation of lipid metabolic process
regulation of MAPK cascade
positive regulation of MAPK cascade
developmental growth
prostate gland growth
epithelial cell differentiation involved in prostate gland development
cellular response to organic substance
negative regulation of hydrogen peroxide-induced cell death
G-protein coupled receptor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-6d74b6fc5b-b8dk7:80/10.239.160.22:80.
git-commit: 0683b6a9b3dadc77a268b704485bdfd9f1fd09b0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.17.0-2024.10.41-1.0