Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ptp4a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011200.2 | NP_035330.1 | 6 | 1566 | ||
GenBank mRNA | AK081491.1 | - | 5 | 1032 | ||
AK148288.1 | - | 6 | 1562 | |||
AK150506.1 | - | 5 | 1196 | |||
AK150556.1 | - | 4 | 2149 | |||
AK150762.1 | - | 4 | 2150 | |||
AK151003.1 | - | 4 | 2150 | |||
AK151533.1 | - | 5 | 1196 | |||
BC003761.1 | - | 6 | 1443 | |||
BC031734.1 | - | 6 | 1488 | |||
BC086787.1 | - | 5 | 872 | 30538771 | ||
BC094447.1 | - | 6 | 1433 | 3990529 | ||
BC150972.1 | - | 5 | 1238 | 9087885 | ||
BC150981.1 | - | 5 | 1241 | 9087894 | ||
U84411.1 | - | 6 | 1575 |