Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ptprs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252453.1 | NP_001239382.1 | 29 | 5151 | ||
NM_001252455.1 | NP_001239384.1 | 28 | 5139 | |||
NM_001252456.1 | NP_001239385.1 | 29 | 5128 | |||
NM_011218.2 | NP_035348.2 | 33 | 6369 | |||
GenBank mRNA | AK144123.1 | - | 21 | 3730 | ||
AK159320.1 | - | 16 | 2432 | |||
AK164645.1 | - | 9 | 1755 | |||
AK169714.1 | - | 12 | 2125 | |||
BC052462.1 | - | 29 | 5077 | 6834684 | ||
BC054077.1 | - | 29 | 5045 | |||
BC083188.1 | - | 28 | 5098 | 5709264 | ||
D28530.1 | - | 33 | 6249 |