Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YES1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005433.3 | NP_005424.1 | 12 | 2610 | ||
XM_005258139.3 | XP_005258196.1 | 11 | 2440 | |||
XM_017025960.1 | XP_016881449.1 | 12 | 2452 | |||
GenBank mRNA | BC048960.1 | - | 12 | 2606 | 5260751 | |
M15990.1 | - | 12 | 2596 |