Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011297.2 | NP_035427.2 | 3 | 179 | ||
NM_207634.1 | NP_997517.1 | 3 | 179 | |||
NM_207635.1 | NP_997518.1 | 3 | 179 | |||
GenBank mRNA | AK002495.1 | - | 3 | 143 | ||
AK002986.1 | - | 542 | ||||
AK005358.1 | - | 3 | 143 | |||
AK007776.1 | - | 3 | 143 | |||
AK008386.1 | - | 3 | 143 | |||
AK009048.1 | - | 3 | 143 | |||
AK010856.1 | - | 3 | 143 | |||
AK012537.1 | - | 3 | 130 | |||
AK018704.1 | - | 3 | 143 | |||
AK049387.1 | - | 139 | ||||
AK052480.1 | - | 3 | 143 | |||
AK135247.1 | - | 3 | 132 | |||
AK151111.1 | - | 3 | 141 | |||
AK167874.1 | - | 2 | 114 | |||
BC081457.1 | - | 3 | 125 | 5712639 | ||
BC086882.1 | - | 3 | 142 | 30120790 | ||
BC091748.1 | - | 3 | 123 | 2812455 | ||
BC092287.1 | - | 2 | 110 | |||
X60289.1 | - | 3 | 134 |