Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scn8a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077499.2 | NP_001070967.1 | 2 | 299 | ||
NM_011323.3 | NP_035453.2 | 1 | 200 | |||
GenBank mRNA | AF049617.1 | - | 1 | 146 | ||
AK158609.1 | - | 2 | 261 | |||
BC099466.1 | - | 2 | 267 | |||
CJ128345.1 | - | 2 | 300 | |||
U26707.1 | - | 1 | 158 |