Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: POU5F2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153216.1 | NP_694948.1 | 1 | 1163 | ||
GenBank mRNA | AK098546.1 | - | 1 | 1163 | ||
BC029532.1 | - | 1 | 1123 | 5295125 | ||
BX641019.1 | - | 1 | 683 |