Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Shcbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011369.2 | NP_035499.1 | 1, 2 | 118 | ||
GenBank mRNA | AF017152.1 | - | 1, 2 | 118 | ||
AK051205.1 | - | 1, 2 | 152 | |||
AK052944.1 | - | 1, 2 | 144 | |||
AK137496.1 | - | 1, 2 | 134 | |||
AK149580.1 | - | 1, 2 | 135 | |||
AK162943.1 | - | 1, 2 | 132 | |||
BC070455.1 | - | 1, 2 | 138 | 30606785 |