Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RTN4 SIRNA Assays ›
Gene Symbol:
RTN4
Gene Name:
reticulon 4
Gene Aliases:
ASY, NI220/250, NOGO, NOGO-A, NOGOC, NSP, NSP-CL, Nbla00271, Nbla10545, Nogo-B, Nogo-C, RTN-X, RTN4-A, RTN4-B1, RTN4-B2, RTN4-C
Chromosome Location:
Chr.2: 54972187 - 55137831 on Build GRCh38
UniGene ID:
Hs.637850
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
6912
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RTN4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001321859.1
NP_001308788.1
9
3590
NM_001321860.1
NP_001308789.1
9
3660
NM_001321861.1
NP_001308790.1
9
3623
NM_001321862.1
NP_001308791.1
9
3610
NM_001321863.1
NP_001308792.1
9
3560
NM_001321904.1
NP_001308833.1
10
4038
NM_007008.2
NP_008939.1
7
1280
NM_020532.4
NP_065393.1
9
4343
NM_153828.2
NP_722550.1
7
1886
NM_207520.1
NP_997403.1
8
1943
NM_207521.1
NP_997404.1
9
3601
NR_135829.1
7
1166
NR_135830.1
7
1144
XM_005264434.3
XP_005264491.1
10
3737
XM_017004518.1
XP_016860007.1
10
3776
XM_017004519.1
XP_016860008.1
10
3893
GenBank mRNA
AF077050.1
-
7
1276
AF087901.1
-
8
1107
AF125103.1
-
7
1280
AF132047.1
-
7
1763
AF132048.1
-
7
1178
AF148537.1
-
10
4122
AF148538.1
-
7
1725
AF177332.1
-
7
1181
AK172726.1
-
5
706
AY102276.1
-
7
1296
AY102277.1
-
7
1828
AY102278.1
-
8
1885
AY102279.1
-
9
4285
AY123245.1
-
9
3598
AY123246.1
-
9
3656
AY123247.1
-
9
3619
AY123248.1
-
9
3605
AY123249.1
-
9
3566
AY123250.1
-
9
3556
BC014366.1
-
7
1174
BC150182.1
-
9
4294
40148210
BC152555.1
-
9
4294
40148984
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
apoptotic process
axonal fasciculation
cerebral cortex radial glia guided migration
negative regulation of cell growth
negative regulation of axon extension
regulation of apoptotic process
negative regulation of axonogenesis
nuclear pore complex assembly
cardiac epithelial to mesenchymal transition
endoplasmic reticulum tubular network organization
endoplasmic reticulum tubular network assembly
cell-cell adhesion
regulation of branching morphogenesis of a nerve
protein binding
poly(A) RNA binding
cadherin binding involved in cell-cell adhesion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-qqtbl:80/10.239.164.61:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0