Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccl11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011330.3 | NP_035460.1 | 3 | 513 | ||
GenBank mRNA | AK009307.1 | - | 3 | 496 | ||
AK010146.1 | - | 3 | 424 | |||
BC027521.1 | - | 3 | 413 | 1527856 | ||
U26426.1 | - | 3 | 412 | |||
U40672.1 | - | 3 | 425 |