Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc2a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011401.4 | NP_035531.3 | 3 | 480 | ||
GenBank mRNA | AK028041.1 | - | 3 | 254 | ||
AK045328.1 | - | 3 | 256 | |||
AK147258.1 | - | 3 | 258 | |||
AK155997.1 | - | 3 | 239 | |||
AK164286.1 | - | 3 | 252 | |||
BC034122.1 | - | 3 | 238 | 4527667 | ||
BC050095.1 | - | 3 | 236 | |||
BC058811.1 | - | 3 | 238 | 6490844 | ||
BQ769655.1 | - | 3 | 482 | |||
CT010354.1 | - | 2 | 125 | |||
M75135.1 | - | 3 | 204 | |||
X61093.1 | - | 3 | 204 |