Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nhp2l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011482.4 | NP_035612.2 | 2, 3 | 229 | ||
GenBank mRNA | AK004489.1 | - | 2, 3 | 177 | ||
AK134943.1 | - | 167 | ||||
AK145391.1 | - | 2, 3 | 231 | |||
AK145608.1 | - | 2, 3 | 211 | |||
AK168808.1 | - | 2, 3 | 199 | |||
AK168930.1 | - | 2, 3 | 182 | |||
AK169103.1 | - | 2, 3 | 213 | |||
BC026755.1 | - | 2, 3 | 175 | 3154066 | ||
BC054450.1 | - | 2, 3 | 175 | 6491298 | ||
BC083315.1 | - | 2, 3 | 151 | 5714819 | ||
U95114.1 | - | 2, 3 | 201 |