Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snai1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011427.2 | NP_035557.1 | 3 | 1206 | ||
GenBank mRNA | AK144478.1 | - | 3 | 1229 | ||
BC034857.1 | - | 3 | 1206 | 5121591 | ||
M95604.1 | - | 3 | 1156 |