Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpinb8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159748.1 | NP_001153220.1 | 2, 3 | 473 | ||
NM_011459.4 | NP_035589.1 | 2, 3 | 473 | |||
GenBank mRNA | AK032659.1 | - | 2, 3 | 473 | ||
AK036811.1 | - | 2, 3 | 409 | |||
AK048230.1 | - | 2, 3 | 433 | |||
AK148413.1 | - | 2, 3 | 471 | |||
BC011076.1 | - | 2, 3 | 452 | |||
BC025121.1 | - | 2, 3 | 463 | 4487039 | ||
U96703.2 | - | 2, 3 | 464 |