Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011484.2 | NP_035614.1 | 2, 3 | 317 | ||
GenBank mRNA | AK035924.1 | - | 2, 3 | 240 | ||
AK144987.1 | - | 2, 3 | 334 | |||
AK147832.1 | - | 2, 3 | 227 | |||
AK157767.1 | - | 2, 3 | 264 | |||
AK162431.1 | - | 2, 3 | 264 | |||
AK163720.1 | - | 2, 3 | 264 | |||
AK163922.1 | - | 2, 3 | 254 | |||
AK169170.1 | - | 2, 3 | 324 | |||
BC044666.1 | - | 2, 3 | 317 | 4950569 | ||
BC055326.1 | - | 2, 3 | 263 | 6416552 | ||
U43900.1 | - | 2, 3 | 174 |