Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srms | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011481.3 | NP_035611.3 | 8 | 2359 | ||
GenBank mRNA | AK049582.1 | - | 8 | 1620 | ||
D26186.1 | - | 8 | 2205 | |||
D49427.1 | - | 8 | 2373 |