Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011588.3 | NP_035718.2 | 2 | 1001 | ||
GenBank mRNA | AF230391.1 | - | 2 | 602 | ||
AF230392.1 | - | 2 | 602 | |||
AK141357.1 | - | 2 | 745 | |||
AK179925.1 | - | 1 | 106 | |||
AK184928.1 | - | 1 | 106 | |||
AK217247.1 | - | 2 | 106 | |||
AK219334.1 | - | 1 | 106 | |||
BC020452.1 | - | 2 | 432 | |||
BC023335.1 | - | 2 | 710 | |||
BC058391.1 | - | 2 | 447 | 6817165 | ||
BC089337.1 | - | 2 | 1001 | 6811294 | ||
U67303.1 | - | 2 | 992 | |||
X99644.1 | - | 2 | 602 |