Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnfrsf9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077508.1 | NP_001070976.1 | 3, 4 | 344 | ||
NM_001077509.1 | NP_001070977.1 | 3, 4 | 325 | |||
NM_011612.2 | NP_035742.1 | 3, 4 | 344 | |||
GenBank mRNA | AK019885.1 | - | 3, 4 | 310 | ||
AK154625.1 | - | 3, 4 | 328 | |||
BC028507.1 | - | 3, 4 | 281 | 1497753 | ||
BY206772.1 | - | 3, 4 | 344 | |||
DQ832278.1 | - | 2, 3 | 230 | |||
DQ832279.1 | - | 2, 3 | 230 |