Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcof1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198984.1 | NP_001185913.1 | 16 | 2490 | ||
NM_011552.3 | NP_035682.1 | 16 | 2490 | |||
GenBank mRNA | AF001794.1 | - | 16 | 2434 | ||
AK089518.1 | - | 16 | 2458 | |||
AK170990.1 | - | 4 | 386 | |||
AK171978.1 | - | 4 | 386 | |||
BC021514.1 | - | 16 | 2495 | 5351783 | ||
BC046608.1 | - | 15 | 2300 | |||
BC052669.1 | - | 16 | 2462 | |||
BC057342.1 | - | 16 | 2443 | 6825735 | ||
BC060105.1 | - | 16 | 2443 | |||
U81030.1 | - | 16 | 2378 |