Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tead3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098226.3 | NP_001091696.2 | 13 | 2161 | ||
NM_001204156.1 | NP_001191085.1 | 13 | 2484 | |||
NM_011566.4 | NP_035696.3 | 13 | 2325 | |||
GenBank mRNA | AF002670.1 | - | 13 | 2280 | ||
AK087649.1 | - | 13 | 2165 | |||
AK143551.1 | - | 12 | 3195 | |||
BC021364.1 | - | 13 | 2205 | |||
BC027383.1 | - | 4 | 1236 | |||
BC039984.1 | - | 4 | 1271 | |||
BC047929.1 | - | 13 | 2484 | |||
D87963.1 | - | 12 | 1995 | |||
Y10027.1 | - | 12 | 2093 |