Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLIC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287593.1 | NP_001274522.1 | 3 | 692 | ||
NM_001287594.1 | NP_001274523.1 | 3 | 232 | |||
NM_001288.4 | NP_001279.2 | 2 | 383 | |||
GenBank mRNA | AF034607.1 | - | 2 | 388 | ||
AF109197.1 | - | 2 | 173 | |||
AK129773.1 | - | 2 | 128 | |||
AK223379.1 | - | 2 | 356 | |||
AK314014.1 | - | 2 | 128 | |||
BC064527.1 | - | 3 | 692 | 5585323 | ||
BC095469.1 | - | 2 | 318 | 30557606 | ||
BM562536.1 | - | 386 | ||||
CR542071.1 | - | 2 | 119 | |||
CX783058.1 | - | 3 | 230 | |||
HY057217.1 | - | 3 | 233 | |||
U93205.1 | - | 2 | 337 | |||
X87689.1 | - | 2 | 323 |