Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039363.2 | NP_001034452.1 | 5 | 543 | ||
NM_001286713.1 | NP_001273642.1 | 5 | 543 | |||
NM_001286714.1 | NP_001273643.1 | 5 | 538 | |||
NM_001286716.1 | NP_001273645.1 | 5 | 538 | |||
NM_001286718.1 | NP_001273647.1 | 5 | 543 | |||
NM_001286720.1 | NP_001273649.1 | 5 | 543 | |||
NM_011647.3 | NP_035777.2 | 5 | 543 | |||
GenBank mRNA | AF132986.1 | - | 1 | 71 | ||
AK143799.1 | - | 5 | 544 | |||
AK147016.1 | - | 5 | 519 | |||
AK147302.1 | - | 5 | 483 | |||
AK147487.1 | - | 5 | 512 | |||
AK147874.1 | - | 5 | 508 | |||
AK147908.1 | - | 5 | 544 | |||
AK148062.1 | - | 5 | 535 | |||
AK148072.1 | - | 5 | 440 | |||
AK170589.1 | - | 5 | 534 | |||
AK171007.1 | - | 5 | 515 | |||
AK190229.1 | - | 2 | 133 | |||
AK214850.1 | - | 2 | 114 | |||
AK217069.1 | - | 2 | 133 | |||
BC052449.1 | - | 5 | 519 | 6409037 | ||
BC060701.1 | - | 5 | 519 | 6409037 | ||
U37775.1 | - | 4 | 458 | |||
U39818.1 | - | 5 | 514 |