Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube2i | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177609.1 | NP_001171080.1 | 5 | 1004 | ||
NM_001177610.1 | NP_001171081.1 | 4 | 293 | |||
NM_011665.4 | NP_035795.1 | 4 | 331 | |||
GenBank mRNA | AK003141.1 | - | 4 | 238 | ||
AK005058.1 | - | 4 | 313 | |||
AK011239.1 | - | 4 | 239 | |||
AK012282.1 | - | 4 | 257 | |||
AK088508.1 | - | 4 | 316 | |||
AK150575.1 | - | 4 | 274 | |||
AK160014.1 | - | 4 | 260 | |||
AK160988.1 | - | 4 | 257 | |||
AK165606.1 | - | 4 | 243 | |||
AK165615.1 | - | 4 | 261 | |||
AK166016.1 | - | 4 | 241 | |||
AK166657.1 | - | 5 | 1005 | |||
AK166930.1 | - | 4 | 237 | |||
AK167162.1 | - | 5 | 474 | |||
AK168212.1 | - | 4 | 260 | |||
AK168706.1 | - | 4 | 220 | |||
AK168766.1 | - | 4 | 254 | |||
AK190279.1 | - | 158 | ||||
AK191598.1 | - | 3 | 150 | |||
AK194165.1 | - | 3 | 150 | |||
BC020437.1 | - | 3 | 179 | |||
BC037635.1 | - | 4 | 223 | 4951933 | ||
BY074880.1 | - | 4 | 333 | |||
U31934.1 | - | 3 | 171 | |||
U76416.1 | - | 4 | 267 | |||
U82627.1 | - | 4 | 276 | |||
U94402.1 | - | 4 | 327 | |||
X97575.1 | - | 4 | 208 | |||
X99739.1 | - | 4 | 255 |