Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011692.2 | NP_035822.2 | 2, 3 | 262 | ||
GenBank mRNA | AK002282.1 | - | 2, 3 | 232 | ||
AK008847.1 | - | 2, 3 | 232 | |||
AK077923.1 | - | 2, 3 | 247 | |||
AK151527.1 | - | 2, 3 | 232 | |||
AK153069.1 | - | 2, 3 | 231 | |||
AK159284.1 | - | 2, 3 | 232 | |||
AK167064.1 | - | 2, 3 | 232 | |||
AK167742.1 | - | 2, 3 | 263 | |||
AK169185.1 | - | 2, 3 | 232 | |||
BC132169.1 | - | 2, 3 | 205 | |||
BC132171.1 | - | 2, 3 | 205 | |||
BY134471.1 | - | 2, 3 | 255 | |||
DV066998.1 | - | 2, 3 | 295 | |||
U96760.1 | - | 2, 3 | 213 |