Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trpv2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011706.2 | NP_035836.2 | 3 | 452 | ||
GenBank mRNA | AB021665.1 | - | 3 | 534 | ||
AK089004.1 | - | 4 | 467 | |||
AK134052.1 | - | 2 | 765 | |||
AK162841.1 | - | 2 | 326 | |||
AK171222.1 | - | 2 | 375 | |||
AK200065.1 | - | 1 | 40 | |||
AK209862.1 | - | 1 | 40 | |||
BB631765.1 | - | 3 | 452 | |||
BC005415.1 | - | 2 | 399 | 3487527 |