Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Baz1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011714.2 | NP_035844.2 | 2 | 501 | ||
GenBank mRNA | AF084480.1 | - | 2 | 460 | ||
AK017894.1 | - | 2 | 502 | |||
AK140172.1 | - | 2 | 289 | |||
AK141305.1 | - | 2 | 242 | |||
BC141399.1 | - | 2 | 404 | 9056012 | ||
BC171975.1 | - | 2 | 404 |