Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tyr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011661.4 | NP_035791.1 | 1 | 346 | ||
GenBank mRNA | AK014619.1 | - | 1 | 386 | ||
AK148172.1 | - | 1 | 369 | |||
AK148341.1 | - | 1 | 296 | |||
AK148357.1 | - | 1 | 369 | |||
AK148432.1 | - | 1 | 325 | |||
AY526904.1 | - | 1 | 387 | |||
BC079678.1 | - | 1 | 296 | 30539846 | ||
D00131.1 | - | 1 | 349 | |||
D00440.1 | - | 1 | 346 | |||
M20234.1 | - | 1 | 447 | |||
M24560.1 | - | 1 | 346 | |||
M26729.1 | - | 1 | 285 | |||
X12782.1 | - | 1 | 387 |