Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube2i | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177609.1 | NP_001171080.1 | 8 | 1864 | ||
NM_001177610.1 | NP_001171081.1 | 7 | 1153 | |||
NM_011665.4 | NP_035795.1 | 7 | 1191 | |||
GenBank mRNA | AK005058.1 | - | 7 | 1173 | ||
AK160014.1 | - | 7 | 1120 | |||
AK160988.1 | - | 7 | 1118 | |||
AK166016.1 | - | 7 | 1101 | |||
AK166930.1 | - | 7 | 1097 | |||
AK168706.1 | - | 7 | 1080 | |||
AK168766.1 | - | 7 | 1114 | |||
AK199807.1 | - | 1 | 124 | |||
U76416.1 | - | 7 | 1144 |