Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trpv2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011706.2 | NP_035836.2 | 6, 7 | 1277 | ||
GenBank mRNA | AB021665.1 | - | 6, 7 | 1359 | ||
AK008689.1 | - | 4, 5 | 802 | |||
AK089004.1 | - | 7, 8 | 1292 | |||
BC005415.1 | - | 5, 6 | 1224 | 3487527 |