Search Thermo Fisher Scientific
Gene Symbol: Map2k5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011840.2 | NP_035970.1 | 1, 2 | 728 | ||
GenBank mRNA | AB019374.1 | - | 1, 2 | 729 | ||
AK020716.1 | - | 1, 2 | 756 | |||
AK159261.1 | - | 1, 2 | 779 | |||
AK166137.1 | - | 1, 2 | 779 | |||
BC013697.1 | - | 1, 2 | 719 | |||
BC028260.1 | - | 1, 2 | 608 | 5067818 |